Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Morpholine for synthesis, Sigma-Aldrich®

Morpholine for synthesis, Sigma-Aldrich®

Flammable
Toxic
Corrosive
Supplier:   Merck
Morpholine for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Morpholine for synthesis, Sigma-Aldrich®

6 Options Available Below

Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Merck Index:12,06362
  • Boiling Pt:128.9 °C (1013 hPa)
  • Melting Pt:–5 °C
  • Density:1 g/cm³ (20 °C)
  • Flash Pt:38 °C
  • Storage Temperature:Ambient
  • UN:2054
  • ADR:8 (3),I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-815-1

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    87.12 g/mol
  • Boiling Point:
    128 - 130 °C (1013 hPa)
  • Vapor Pressure:
    10 hPa (20 °C)
  • Flash Point:
    32 °C
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    10.6 (5 g/l, H2O, 20 °C)
  • Ignition Point:
    275 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 0,1 L
    1 * 0,1 L - 32,00 €
    32,00 €
    1 * 0,1 L
       32,00 €

  • Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 47,40 €
    47,40 €
    1 * 500 mL
       47,40 €

  • Plastic bottle for solids
    10 kg
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 67,90 €
    67,90 €
    1 * 1 L
       67,90 €

  • Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 155,00 €
    155,00 €
    1 * 2,5 L
       155,00 €

  • Receptacle PE/metal, returnable
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.