Morpholine 99+%, extra pure
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:110-91-8
- Formula:C₄H₉NO
- MDL Number:MFCD00005972
- Molecular Weight:87.12 g/mol
- Merck Index:12,06362
- Boiling Pt:126…130 °C (760 mmHg)
- Melting Pt:–5 °C
- Density:0.99 g/cm³
- Flash Pt:32 °C
- Storage Temperature:Ambient
- EINECS:203-815-1
- UN:2054
- ADR:8,I
4 Options Available Below
Product Details & Documents
- CAS Number:110-91-8
- Formula:C₄H₉NO
- MDL Number:MFCD00005972
- Molecular Weight:87.12 g/mol
- Merck Index:12,06362
- Boiling Pt:126…130 °C (760 mmHg)
- Melting Pt:–5 °C
- Density:0.99 g/cm³
- Flash Pt:32 °C
- Storage Temperature:Ambient
- EINECS:203-815-1
- UN:2054
- ADR:8,I
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤20 APHA
- Insoluble matter:≤0.01 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



