Cycloheximide
Glass bottle for solids, 1 g
Catalog # SANTSC-3508
Marketsource
Supplier: Santa Cruz Biotechnology
Cycloheximide
Catalog # SANTSC-3508
Supplier: Santa Cruz Biotechnology
Marketsource
CAS Number: 66-81-9
Jump to:
Product Details & Documents
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35 g/mol
- Storage Temperature:Ambient
- EINECS:200-636-0
- UN:2811
- ADR:6.1,I
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



