
2-Piperidone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:675-20-7
:
Formula:C₅H₉NO
MDL Number:MFCD00006037
:99.13 g/mol
Boiling Pt:256…262 °C (760 mmHg)
Melting Pt:35…39 °C
Density:1.073 g/cm³
Flash Pt:147 °C
EINECS:211-622-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to brown Low melting crystalline mass
- Infrared spectrum:Conforms
- GC:≥98.5 %
- HPLC:≥96.0 % as diethylsulfonamide
Specification Test Results
Frequently Bought Together