Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Hydroxycinnamic acid
4-Hydroxycinnamic acid
4-Hydroxycinnamic acid
ROTH9908.1
:
Roth Carl
:
4-Hydroxycinnamic acid
ROTH9908.1
:
Roth Carl
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
164.16 g/mol
:
231-000-0
:
GHS07
:
7400-08-0
:
501-98-4
:
C₉H₈O₃
:
MFCD00004399