
trans-4-Hydroxycinnamic acid 98%
Leverancier: Thermo Fisher Scientific
Certificaten
Over dit artikel
CAS-nummer:501-98-4
:7400-08-0
Formule:C₉H₈O₃
MDL-nummer:MFCD00004399
:164.16 g/mol
Smelten Pt:214 °C (dec.)
EINECS:231-000-0
:GHS07
Specificatie testresultaten
- Appearance:White to yellow to beige to green to light brown Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- GC:≥95.0 %
- Stabilizer:200 to 300 ppm MEHQ
Specificatie testresultaten
Frequently Bought Together