Order Entry
Netherlands
ContactUsLinkComponent
Maleic anhydride ≥98%, TECHNICAL
Irritant
Corrosive
Health
Maleic anhydride ≥98%, TECHNICAL
Catalog # 21409.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Health
Maleic anhydride ≥98%, TECHNICAL
Catalog # 21409.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :98.06 g/mol
Merck Index:13,05727
Boiling Pt:200 °C (1013 hPa)
Melting Pt:51…53 °C
Density:1.48 g/cm³ (20 °C)
Flash Pt:103 °C
EINECS:203-571-6
UN:2215
ADR:8,III
 :GHS07
 :GHS05
 :GHS08
Storage Temperature:Ambient
CAS Number:108-31-6
Formula:C₄H₂O₃
MDL Number:MFCD00005518