Fumaric acid NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:110-17-8
:
Formula:C₄H₄O₄
MDL Number:MFCD00002700
:116.07 g/mol
Merck Index:13,04308
Boiling Pt:356 °C (1013 hPa)
Melting Pt:287 °C
Density:1.625 g/cm³ (20 °C)
Flash Pt:273 °C
EINECS:203-743-0
:GHS07
Related Information
Frequently Bought Together