Order Entry
Netherlands
ContactUsLinkComponent
Dibutyl maleate for synthesis, Sigma-Aldrich®
Health
Irritant
Environment
Dibutyl maleate for synthesis, Sigma-Aldrich®
Catalog # 8.18688.9050
Supplier:  Merck
CAS Number:  
Health
Irritant
Environment
Dibutyl maleate for synthesis, Sigma-Aldrich®
Catalog # 8.18688.9050
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • Solubility:
    0.173 g/l (20 °C)
  • Melting Point:
    -85 °C
  • Molar Mass:
    228.29 g/mol
  • Boiling Point:
    280 - 282 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    120 °C
  • Refractive Index:
    1.4451 (20 °C, 589 nm)
  • Explosion Limit:
    0.5 - 3.4 %(V)
  • Density:
    0.99 g/cm3 (20 °C)
  • Ignition Point:
    280 °C

Specification Test Results

About this item

 :
 :228.29 g/mol
Boiling Pt:280 °C (1013 hPa)
Density:0.9938 g/cm³ (20 °C)
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:203-328-4
UN:3082
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:105-76-0
Formula:C₁₂H₂₀O₄
MDL Number:MFCD00009447