Order Entry
Netherlands
ContactUsLinkComponent
cis-Dichloroethylene
cis-Dichloroethylene
  48597.
 :  Merck
 :  
undefined
cis-Dichloroethylene
  48597.
 :  Merck
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :96.94 g/mol
 :60 °C (1013 hPa)
 :9.5 °C
 :1.282 g/cm³ (20 °C)
 :Refrigerator
 :205-859-7
 :1150
 :3,II
 :GHS02
 :GHS07
 :GHS09
 :156-59-2
 :C₂H₂Cl₂
 :MFCD00000925