Order Entry
Netherlands
ContactUsLinkComponent
 
Sodium glycodeoxycholate ≥96% (by TLC), Millipore®
  361311-5
 :  
Sodium glycodeoxycholate ≥96% (by TLC), Millipore®
  361311-5
 :  

 

  • Pack type:
    Plastic ampoule
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Purity (TLC):
    ≥96%
  • pKa:
    4.8
  • Aggregation Number:
    2
  • CMC:
    2.1 mM

 

 

 :
 :471.60 g/mol
 :Ambient
 :16409-34-0
 :C₂₆H₄₂NNaO₅
 :MFCD00036742