Order Entry
Netherlands
ContactUsLinkComponent
 
tert-Butyl methyl ether for HPLC
 
undefined
tert-Butyl methyl ether for HPLC

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.80 %
  • UV:
    1 cm cell vs HPLC-grade water
  • Residue after evaporation:
    ≤0.0003 %
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g

 

 

Frequently Bought Together