Order Entry
Netherlands
ContactUsLinkComponent
AMP (2-Amino-2-methylpropanol) for synthesis, Sigma-Aldrich®
AMP (2-Amino-2-methylpropanol) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
AMP (2-Amino-2-methylpropanol) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    30 - 31 °C
  • Molar Mass:
    89.14 g/mol
  • Boiling Point:
    165 °C (1013 hPa) (anhydrous)
  • Vapor Pressure:
    0.45 hPa (20 °C)
  • Flash Point:
    77.8 °C
  • Refractive Index:
    1.448 (20 °C, 589 nm)
  • Density:
    0.95 g/cm3 (25 °C)
  • pH:
    11.3 (10 g/l, H2O)
  • Ignition Point:
    438 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together