Order Entry
Netherlands
ContactUsLinkComponent
 
Isobutyric acid for synthesis, Sigma-Aldrich®
 :  Merck
 
Isobutyric acid for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    210 g/l (20 °C)
  • Melting Point:
    -45 °C
  • Molar Mass:
    88.11 g/mol
  • Boiling Point:
    153 - 155 °C (1013 hPa)
  • Vapor Pressure:
    12 hPa (20 °C)
  • Flash Point:
    57.5 °C
  • Refractive Index:
    1.3925 (20 °C, 589 nm)
  • Explosion Limit:
    2 - 12 %(V)
  • Density:
    0.95 g/cm3 (20 °C)
  • Ignition Point:
    420 °C

 

Frequently Bought Together