


Acetic anhydride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-24-7
:
Formula:(CH₃CO)₂O
MDL Number:MFCD00008705
Merck Index:14,56
Boiling Pt:138…140 °C
Melting Pt:–73 °C
Density:1.082
Flash Pt:49 °C (120 °F)
UN:1715
ADR:8,II
:102.09 g/mol
Storage Temperature:Ambient
EINECS:203-564-8
REACH:01-2119486470-36
:GHS06
:GHS05
:GHS02
Related Information
Frequently Bought Together