
Bis(2-ethoxyethyl)ether 99+% for HPLC
Catalogus # 43464.AK
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Seq:2590
Specifications
About this item
:
:162.23 g/mol
Merck Index:14,3118
Boiling Pt:188…190 °C
Melting Pt:–44 °C
Density:0.909
Flash Pt:71 °C (160 °F)
Storage Temperature:Ambient
EINECS:203-963-7
:GHS07
CAS Number:112-36-7
Formula:C₈H₁₈O₃
MDL Number:MFCD00009254