2-(Dimethylamino)-ethanol
Supplier: Serva
Certificates
About this item
CAS Number:108-01-0
:
Formula:C₄H₁₁NO
MDL Number:MFCD00002846
:89.14 g/mol
Merck Index:13,02863
Boiling Pt:132…135 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.8871 g/cm³ (20 °C)
Flash Pt:39 °C
Storage Temperature:Ambient
EINECS:203-542-8
UN:2051
ADR:8,II
:GHS02
:GHS05
:GHS07
Frequently Bought Together