2-(Dimethylamino)-ethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-01-0
:
Formula:C₄H₁₁NO
MDL Number:MFCD00002846
:89.14 g/mol
Merck Index:13,02863
Boiling Pt:139 °C (760 mmHg)
Melting Pt:–70 °C
Density:0.888 g/cm³
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:203-542-8
UN:2051
ADR:8,II
:GHS06
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4290 to 1.4300 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
- Specific gravity:(20°C) 0.885 to 0.889
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0006 meq/g
- Aldehyde (HCHO):≤0.002 %
- Methanol:≤0.05 %
- KMnO4-reducing substance:Passes test
Specification Test Results
Frequently Bought Together