


N,N-Dimethyloctylamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7378-99-6
:
Formula:C₁₀H₂₃N
MDL Number:MFCD00009558
:157.3 g/mol
Boiling Pt:195 °C (760 mmHg)
Melting Pt:–57 °C
Density:0.765 g/cm³
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:230-939-3
UN:2922
ADR:8,II
:GHS06
:GHS05
:GHS09
Specification Test Results
- Appearance:Clear to slightly hazy liquid
- Infrared spectrum:Conforms
- Refractive index:1.4233 to 1.4253 (20°C, 589 nm)
- GC:≥96.0 % (C8-chains)
- Color scale:≤30 APHA
- Titration with HClO4:≥96.0 %
Specification Test Results
Frequently Bought Together