Thioacetamide ACS, Reag. Ph. Eur., GR for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:62-55-5
:
Formula:CH₃CSNH₂
MDL Number:MFCD00008070
:75.13 g/mol
Merck Index:13,09391
Melting Pt:112…114 °C
Density:1.37 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:200-541-4
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (argentometric):≥ 99.0 %
- Identity (IR-spectrum):passes test
- Melting point:111 - 114 °C
- Heavy metals (as Pb):≤ 0.001 %
- Fe (Iron):≤ 0.0005 %
- Sulfated ash:≤ 0.1 %
Specification Test Results
Related Information
Frequently Bought Together