Triammonium citrate ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97+% CAS No.: 3458-72-8 Molecular Formula: C6H17N3O7 Formula Weight: 243.22 Physical Form: Crystalline powder Appearance: White Melting Point: ca 180° dec. Solubility: Soluble in water MDL No.: MFCD00036406
Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate
- CAS Number:3458-72-8
- Formula:C₆H₁₇N₃O₇
- MDL Number:MFCD00036406
- Molecular Weight:243.22 g/mol
- Melting Pt:∼ 180 °C (dec.)
- Density:1 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:222-394-5
2 Options Available Below
Product Details & Documents
Purity: 97+% CAS No.: 3458-72-8 Molecular Formula: C6H17N3O7 Formula Weight: 243.22 Physical Form: Crystalline powder Appearance: White Melting Point: ca 180° dec. Solubility: Soluble in water MDL No.: MFCD00036406
Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate
- CAS Number:3458-72-8
- Formula:C₆H₁₇N₃O₇
- MDL Number:MFCD00036406
- Molecular Weight:243.22 g/mol
- Melting Pt:∼ 180 °C (dec.)
- Density:1 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:222-394-5
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



