Ammonium formate ≥97%, AnalaR NORMAPUR® analytical reagent
Catalog # 21254.260
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay:Min. 97 %
- Identification:Passes test
- pH (20°C; 5 %):6 - 7
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 100 ppm
- SO4 (Sulphate):Max. 100 ppm
- Fe (Iron):Max. 20 ppm
Specification Test Results
About this item
:
:63.06 g/mol
Merck Index:13,00525
Melting Pt:119…121 °C
Density:1.28 g/cm³ (25 °C)
EINECS:208-753-9
:GHS07
Storage Temperature:Ambient
CAS Number:540-69-2
Formula:HCO₂NH₄
MDL Number:MFCD00013103