Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Barium acetate
Barium acetate ≥99%
Barium acetate ≥99%
A10316.30
:
Thermo Fisher Scientific
:
Barium acetate ≥99%
A10316.30
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
250 g
Seq:
3545
:
543-80-6
:
:
(H₃CCOO)₂Ba
:
MFCD00012447
:
255.42 g/mol
:
14,965
:
∼450 °C
:
2.47
:
Ambient
:
208-849-0
:
1564
:
6.1,III
:
GHS07