
1-Naphthyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:830-81-9
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00003922
:186.21 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:43…46 °C
Density:1.1305 g/cm³ (50 °C)
Flash Pt:>110 °C (230 °F)
EINECS:212-599-8
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together