Order Entry
Netherlands
ContactUsLinkComponent
1-Naphthyl acetate 99%
1-Naphthyl acetate 99%
Catalog # ACRO227250250
CAS Number:  
undefined
1-Naphthyl acetate 99%
Catalog # ACRO227250250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to light yellow to light pink Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    42 to 47 °C
  • HPLC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :186.21 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:43…46 °C
Density:1,1305 g/cm³ (50 °C)
Flash Pt:≥ 100 °C
Storage Temperature:Refrigerator
EINECS:212-599-8
CAS Number:830-81-9
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00003922