Hydroxyurea
Glass bottle for solids, 5 g
SANTSC-29061
Marketsource
: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- :127-07-1
- :CH₄N₂O₂
- :MFCD00007943
- :76.05 g/mol
- :13,04874
- :296 °C (1013 hPa)
- :142…144 °C
- :Ambient
- :204-821-7
- Pack type:Glass bottle for solids
- Pk:5 g