

Methyl pyruvate 94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:600-22-6
:
Formula:C₄H₆O₃
MDL Number:MFCD00008754
:102.09 g/mol
Boiling Pt:134…137 °C (760 mmHg)
Density:1.13 g/cm³
Flash Pt:39 °C
Storage Temperature:Refrigerator
EINECS:209-987-4
UN:3272
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4035 to 1.4075 (20°C, 589 nm)
- GC:≥92 %
Specification Test Results
Frequently Bought Together