Order Entry
Netherlands
ContactUsLinkComponent
Chloral hydrate
Toxic
Chloral hydrate
Catalog # EHERC11098000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Toxic
Chloral hydrate
Catalog # EHERC11098000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :165.4 g/mol
Merck Index:12,02113
Boiling Pt:97.5 °C (1013 hPa)
Melting Pt:52 °C
Density:1.901 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-117-5
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:302-17-0
Formula:Cl₃CCH(OH)₂
MDL Number:MFCD00044479