Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,2-Dibromo-3-Chloropropane Neat Standar
Supelco® Organic Reference Standard, 1,2-Dibromo-3-Chloropropane Neat Standar
Catalog # 395587-5X1G
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 1,2-Dibromo-3-Chloropropane Neat Standar
Catalog # 395587-5X1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Description:
    1,2-DIBROMO-3-CHLOROPROPANE NEAT STANDAR
  • Storage conditions:
    +2...+8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item