Potassium sorbate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:24634-61-5
:590-00-1
Formula:CH₃CH=CHCH=CHCOOK
MDL Number:MFCD00016546
:150.22 g/mol
Merck Index:14,7671
Melting Pt:>300 °C
Density:1.361
EINECS:246-376-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together