Order Entry
Netherlands
ContactUsLinkComponent
Sorbic acid
Sorbic acid
Catalog # EHERC16971500
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Sorbic acid
Catalog # EHERC16971500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :112.13 g/mol
Merck Index:13,08793
Boiling Pt:228 °C (1013 hPa)
Melting Pt:132…135 °C
Density:1.204 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-768-7
 :GHS07
CAS Number:110-44-1
Formula:CH₃CH=CHCH=CHCOOH
MDL Number:MFCD00002703