Methyl linoleate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-63-0
:
Formula:C₁₉H₃₄O₂
MDL Number:MFCD00009534
:294.48 g/mol
Merck Index:14,06095
Boiling Pt:346°C
Melting Pt:–35°C
Density:0,89 g/cm³ (20 °C)
Flash Pt:200°C
EINECS:203-993-0
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together