Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Biotin
D-(+)-Biotin
D-(+)-Biotin
D-(+)-Biotin
SANBMONX10959
:
Hycultec
:
D-(+)-Biotin
SANBMONX10959
:
Hycultec
:
Pack type:
Glass bottle
Pk:
1 mL
Seq:
2070
:
:
244.31 g/mol
:
13,01231
:
232…233 °C
:
1.41 g/cm³ (20 °C)
:
Refrigerator
:
200-399-3
:
58-85-5
:
C₁₀H₁₆N₂O₃S
:
MFCD00005541