D-(-)-Histidine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 351-50-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: ca 285° dec. Beilstein Reference: 84089
Synonyms: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
- CAS Number:351-50-8
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00065963
- Molecular Weight:155.16 g/mol
- Merck Index:12,04758
- Melting Pt:∼ 285 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-513-8
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 351-50-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: ca 285° dec. Beilstein Reference: 84089
Synonyms: (R)-(-)-Histidine, D-3-Imidazol-4-ylalanine, (2R)-(-)-Histidine, H-D-His-OH
- CAS Number:351-50-8
- Formula:C₆H₉N₃O₂
- MDL Number:MFCD00065963
- Molecular Weight:155.16 g/mol
- Merck Index:12,04758
- Melting Pt:∼ 285 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-513-8



