D-(-)-Mannitol
Catalog # DIFC217020
Supplier: DIFCO BD
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
:
:182.17 g/mol
Boiling Pt:290…295 °C (4 hPa)
Melting Pt:164…169 °C
Density:1.49 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-711-8
CAS Number:69-65-8
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287
Merck Index:13,05769