
(S)-(+)-1-Aminopropan-2-ol 98+%
:2799-17-9
:
:C₃H₉NO
:MFCD00064429
:75.11 g/mol
:160 °C (760 mmHg)
:19…21 °C
:0.954 g/cm³
:71 °C
:Ambient
:220-533-4
:2735
:8,III
:GHS05
- Appearance:Clear colorless to light yellow Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.4465 to 1.4495 (20°C, 589 nm)
- GC:≥98.0 %
- Specific optical rotation:+16° to +20° (20°C, 589 nm) (c=1.8, H2O)
Frequently Bought Together