Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020300IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020300IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    52663-76-0
  • Description:
    2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₂Cl₈
  • Description:
    2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203)

Specifications

About this item