Order Entry
Netherlands
ContactUsLinkComponent
 
Organic reference standard, 2,3,4-Trichlorobiphenyl (PCB No. 21) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20002100IO
 :  
Organic reference standard, 2,3,4-Trichlorobiphenyl (PCB No. 21) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20002100IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    55702-46-0
  • Description:
    2,3,4-Trichlorobiphenyl (PCB No. 21) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,3,4-Trichlorobiphenyl (PCB No. 21)