Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

D-(-)-Glutamic acid ≥99%

D-(-)-Glutamic acid ≥99%
Certificates
D-(-)-Glutamic acid ≥99%
D-(-)-Glutamic acid ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 6893-26-1 Molecular Formula: C5H9NO4 Formula Weight: 147.13 Melting Point: 199° dec. Beilstein Reference: 1723800 Merck Reference: 14,4469 Optical Rotation: -30° (c=10 in 2N HCl) MDL No.: MFCD00063112

Synonyms:  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  • CAS Number:6893-26-1
  • Formula:C₅H₉NO₄
  • MDL Number:MFCD00063112
  • Molecular Weight:147.13 g/mol
  • Merck Index:14,4469
  • Boiling Pt:325 °C (1013 hPa)
  • Melting Pt:199 °C (dec.)
  • Density:1.533 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:230-000-8
Product Family Options

Product Information