Organic reference standard, 2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20011800IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:31508-00-6
- Description:2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3',4,4',5-Pentachlorobiphenyl (PCB No. 118)