Organic reference standard, 2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20010300IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:60145-21-3
- Description:2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103)