Supelco® Organic Reference Standard, PCB No 101 Solution
Catalog # 36902-2ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:37680-73-2
- Description:PCB No 101 solution
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055553
- M-Clarity quality level:MQ100
- Synonym(s):2,2′,4,5,5′-Pentachlorobiphenyl solution; 2,2′,4,5,5′-PCB
- Formula:C₁₂H₅Cl₅
- Molecular weight:326,43
- Flash point:4 °C - closed cup
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 + P331 - P302 + P352