
L(+)-Lactic acid 90% in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-33-4
:
Formula:C₃H₆O₃
MDL Number:MFCD00064266
:90.08 g/mol
Merck Index:13,05352
Boiling Pt:125 °C
Melting Pt:~ 53 °C
Density:1.2 g/cm³
Flash Pt:>112 °C
EINECS:201-196-2
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4260 to 1.4280 (20°C, 589 nm)
- Sulfated ash:≤0.1 %
- Density:(20°C) 1.2 to 1.21 g/ml
- Identification:Passes test
- Titration excess NaOH:≥88 %
- Chloride (Cl):≤0.2 %
- Sulfate (SO4):≤0.25 %
- Arsenic (As):≤3 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Sugars:Passes test
Specification Test Results
Frequently Bought Together