Order Entry
Netherlands
Orders LinkContactUsLinkComponent
D(-)-Lactic acid

D(-)-Lactic acid

Corrosive
Marketsource
Supplier:   BACHEM BIOCHEMICA
D(-)-Lactic acid
Supplier:  BACHEM BIOCHEMICA
Marketsource
Certificates
D(-)-Lactic acid

2 Options Available Below

Product Details & Documents
Synonyms:  (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
  • CAS Number:10326-41-7
  • Formula:C₃H₆O₃
  • MDL Number:MFCD00068313
  • Molecular Weight:90.08 g/mol
  • Storage Temperature:Freezer
  • EINECS:233-713-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.