Organic reference standard, 2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20004700IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2437-79-8
- Description:2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,2',4,4'-Tetrachlorobiphenyl (PCB No. 47