Organic reference standard, 3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalogus # EHERL20007700IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:32598-13-3
- Description:3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77) 10 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77