Organic reference standard, 2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20016700IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:52663-72-6
- Description:2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167) 10 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)