Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20018300IO
 :  
Organic reference standard, 2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183) 10 µg/ml in isooctane, Dr Ehrenstorfer
  EHERL20018300IO
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    52663-69-1
  • Description:
    2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,2',3,4,4',5',6-Heptachlorobiphenyl (PCB No. 183)