D-(-)-Glutamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5959-95-5
:
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00065607
:146.15 g/mol
:GHS07
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Specific optical rotation:-30° to -34° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together