N-Acetyl-L-cysteine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:616-91-1
:
Formula:C₅H₉NO₃S
MDL Number:MFCD00004880
:163.2 g/mol
Merck Index:14,88
Boiling Pt:371 °C (1013 hPa)
Melting Pt:108…111 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-498-3
Related Information
Frequently Bought Together